Learn More
Diagnostic Biosystems CYTOKERATIN 15 Cytokeratin 15
Supplier:  Diagnostic Biosystems MOB30505
Encompass_Preferred
Cytokeratin 15 is a type I cytokeratin without a defined type II partner. Cytokeratin 15 is expressed primarily in the basal keratinocytes of stratified tissues, including the fetal epidermis and fetal nail.
Catalog No. 50-238-5450
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.