Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 15 Monoclonal Antibody (LKH15)
GREENER_CHOICE

Catalog No. PIMA190926 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA190926 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA190926 Supplier Invitrogen™ Supplier No. MA190926
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-90926 detects Cytokeratin 15 from human samples.

Cytokeratin 15 is a member of the keratin gene family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. Most of the type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains and are clustered in a region on chromosome 17q21. 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 15
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LKH15
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide
Gene KRT15
Gene Accession No. P19012
Gene Alias AI528832; C K1CO; CK15; CK-15; cytokeratin 15; cytokeratin-15; K15; K1CO; Ka15; keratin 15; keratin 15, type I; keratin complex 1, acidic, gene 15; keratin, type I cytoskeletal 15; Keratin-15; keratin-15, basic; keratin-15, beta; KRT1; Krt1-15; KRT15; krt15 {ECO:0000250; KRTB; type I cytoskeletal 15; type I keratin KA15; UniProtKB:P19012}
Gene Symbols KRT15
Host Species Mouse
Immunogen A 17-mer synthetic peptide from the C-terminus of human cytokeratin 15.
Purification Method IgG fraction
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3866
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.