Learn More
Invitrogen™ Cytokeratin 19 Monoclonal Antibody (A53-B/A2), Biotin

Description
This antibody recognizes the rod domain of cytokeratin 19 (40 kDa), an intracellular antigen constituting intermediate cytoskeleton filaments. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7.
Specifications
Specifications
| Antigen | Cytokeratin 19 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A53-B/A2 |
| Concentration | 1 mg/mL |
| Conjugate | Biotin |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | KRT19 |
| Gene Accession No. | P08727 |
| Gene Alias | 40-kDa keratin intermediate filament; AI663979; C K1CS; CK 19; CK19; CK-19; cytokeratin 19; cytokeratin-19; EndoC; K19; K1CS; Ka19; keratin 19; keratin 19, type I; keratin complex 1, acidic, gene 19; keratin, type I cytoskeletal 19; keratin, type I, 40-kd; keratin-19; Krt-1.19; Krt1-19; Krt19; LOC100344434; MGC15366; Type I keratin Ka19 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.