Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 3 Monoclonal Antibody (AE5)
GREENER_CHOICE

Catalog No. 01675182
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-182 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-182 Supplier Invitrogen™ Supplier No. MA15763
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-5763 detects the 64k basic keratin K3 in human, rabbit, rat and bovine samples. MA1-5763 has been successfully used in Western blot, immunocytochemistry/immunofluorescence and immunohistochemistry (paraffin) procedures. The MA1-5763 immunogen is Rabbit corneal epithelial keratins.

Cytokeratin 3 is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the corneal epithelium with family member KRT12 and mutations in these genes have been associated with Meesmann's Corneal Dystrophy. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. (NCBI Gene description).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot
Classification Monoclonal
Clone AE5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene KRT3
Gene Accession No. P12035, Q29426
Gene Alias 65 kDa cytokeratin; CK3; CK-3; cytokeratin 3; cytokeratin type II, component Ib/c; cytokeratin-3; K3; K3 keratin; keratin 3; keratin 3, type II; keratin K3; keratin, type II cytoskeletal 3; Keratin-3; KERIB; KRT3; Type-II keratin Kb3
Gene Symbols KRT3
Host Species Mouse
Immunogen Rabbit corneal epithelial keratins from P3 x 63 Ag8 cells.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100353668, 3850, 407188
Target Species Human, Rabbit, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.