Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 37/38 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106801 Supplier Invitrogen™ Supplier No. PA5106801
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total KRT37/38.

The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 37/38
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT37
Gene Accession No. O76014, O76015
Gene Alias HA7; HA8; hair keratin; hair keratin, type I Ha7; Hair keratin, type I Ha8; HHA7; HHA8; HKA7; HKA8; K37; K38; keratin 37; keratin 37, type I; keratin 38; keratin 38, type I; keratin, hair, acidic, 7; keratin, hair, acidic, 8; keratin, type I cuticular Ha7; Keratin, type I cuticular Ha8; keratin-37; Keratin-38; KRT hHa8; KRT37; KRT38; KRTHA7; KRTHA8
Gene Symbols KRT37, KRT38
Host Species Rabbit
Immunogen A synthesized peptide derived from human KRT37(Accession O76014), corresponding to amino acid residues Y355-R405.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8687, 8688
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.