Learn More
Biocare Medical, LLC Cytokeratin 5/6 Antibody - Concentrate, 1.0 ml
SDP
Supplier:  Biocare Medical, LLC CM105C
Encompass_Preferred
Studies have shown Cytokeratin 5/6 antibody reacts with human epidermis and non-keratinizing epithelium. It has also been shown to react with cytokeratin 6, weakly with cytokeratin 4, and does not react with cytokeratins 1, 7, 8, 10, 13, 14, 18 and 19. CK5/6 has been shown to express in the vast majority of squamous cell carcinoma, basal cell carcinomas, thymomas, salivary gland tumors, and mesothelioma. It rarely reacts with pulmonary adenocarcinomas.
Catalog No. 50-825-15
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.