Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 5/8 Monoclonal Antibody (C50)
GREENER_CHOICE

Catalog No. 01674359
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-674-359 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-674-359 Supplier Invitrogen™ Supplier No. MA112591
  • $599.00
    $497.30 / Each of 1

    Save $101.70

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Cytokeratin 5/8 Monoclonal Antibody for Western Blot, IHC (F)

Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between ~40 kDa and ~68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 5/8
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone C50
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide
Gene KRT5
Gene Accession No. P05787, P13647
Gene Alias 3300001P10Rik; 58 kDa cytokeratin; AA960620; AL022697; AU019895; AW146334; Card2; CK5; CK-5; CK8; CK-8; CYK8; CYKER; cytokeratin 5; cytokeratin 8; cytokeratin 8 (370 AA); cytokeratin endo A; cytokeratin type II, component III; cytokeratin-5; Cytokeratin-8; cytokeratin-A; DDD; DDD1; EBS2; EndoA; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K2C8; K5; K8; KERA; keratin 5; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin 8; keratin 8, type II; keratin complex 2, basic, gene 5; keratin complex 2, basic, gene 8; keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 60 kDa, component III; Keratin, type II cytoskeletal 8; keratin, type II cytoskeletal 8-like protein; keratin-5; Keratin-8; KO; Krt-2.8; Krt2-5; Krt2-8; krt5; KRT5A; Krt8; Sak57; Tfip8; tuftelin interacting protein 8; type II cytokeratin A (no.8); type-II keratin Kb5; Type-II keratin Kb8
Gene Symbols KRT5, KRT8
Host Species Mouse
Immunogen Cytoskeleton preparation of HeLa Cells.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3852, 3856
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.