Learn More
Biocare Medical, LLC Cytokeratin 5 Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM234A
Encompass_Preferred
Cytokeratin 5 Antibody is a 58 kDa protein that is closely related to CK6. ELISA testing has shown the XM26 clone was positive for CK5 and negative for the CK6 protein. CK5 is in many non-keratinizing stratified squamous epithelia such as tongue mucosa, basal epithelia hair follicles and trachea. It is also expressed in prostate gland basal cells, mammary gland myoepithelial cells and most epithelial and biphasic mesotheliomas. According to various studies, CK5 is expressed in large cell carcinomas and pulmonary squamous cell carcinomas. The sensitivity of CK5 for identifying basal-like tumors in breast was 97% compared with 59% for CK5/6.
Catalog No. 50-826-18
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.