Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 5 Monoclonal Antibody (GT1486)
GREENER_CHOICE

Catalog No. PIMA531581
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531581 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531581 Supplier Invitrogen™ Supplier No. MA531581
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (97%), Rat (97%), Dog (97%), Chicken (80%), Chimpanzee (100%), Bovine (97%). Positive Control: HCT-116. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytokeratin pan is part of a subfamily of intermediate filament proteins that are characterized by remarkable biochemical diversity, and represented in human epithelial tissues by at least 20 different polypeptides. Cytokeratins range in molecular weight between 40 kDa- 68 kDa, and an isoelectric pH between 4.9-7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays. The composition of cytokeratin pairs vary with the epithelial cell type, stage of differentiation, cellular growth environment, and disease state. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone GT1486
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene KRT5
Gene Accession No. P13647, Q6P6Q2, Q922U2
Gene Alias 3300001P10Rik; 58 kDa cytokeratin; alpha keratin; AW146334; CK5; CK-5; ckii; ckii protein; cytokeratin 5; cytokeratin-5; DDD; DDD1; EBS2; epidermolysis bullosa simplex 2 Dowling-Meara/Kobner/Weber-Cockayne types; K5; keratin 5; keratin 5 (epidermolysis bullosa simplex Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5 (epidermolysis bullosa simplex, Dowling-Meara/Kobner/Weber-Cockayne types); keratin 5, type II; keratin complex 2, basic, gene 5; keratin type II, tail domain; keratin, type II cytoskeletal 5; keratin, type II cytoskeletal 5; keratin 5; keratin-5; Krt2-5; KRT5; KRT5A; Sak57; Tfip8; tuftelin interacting protein 8; type II alpha-keratin IIA; type II cytokeratin; type-II keratin Kb5; wu:fb02h09; wu:fj02d02; wu:fk78b12; zfCKII
Gene Symbols KRT5
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human Cytokeratin 5. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 110308, 369017, 3852
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.