Learn More
Diagnostic Biosystems CYTOKERATIN 5AND14 Cytokeratin 5/14
Supplier:  Diagnostic Biosystems PDM140
Encompass_Preferred
This antibody cocktail reacts with cytokeratin 5 intermediate filament protein as well as the 50 kDa cytokeratin protein designated cytokeratin 14. Basal cells of human epidermis express acidic cytokeratin 14 and basic cytokeratin 5. Cytokeratin 5 is a 58 kDa protein, which is closely related to cytokeratin 6. Cytokeratin 5 and 6 are expressed in basal cell epithelioma, basal cells of prostate, urothelium, vagina, squamous cell carcinomas ofskin, tongue, epiglottis and rectal-anal region. Cytokeratin 5 can be useful in the distinction of mesotheliomas from most adenocarcinomas. Cytokeratin 14 belongs to the type A (acidic) subfamily of low MW cytokeratins and existsin combination with cytokeratin 15. This antibody can be useful to distinguish stratified epithelial cellsfrom simple epithelial cells.
Catalog No. 50-238-5542
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.