Learn More
Diagnostic Biosystems CYTOKERATIN 7 Cytokeratin 7 (LP1K)
Supplier:  Diagnostic Biosystems MOB56301
Encompass_Preferred
Keratins are cytoplasmic intermediate filament proteins expressed by epithelial cells. Cytokeratin 7 (CK7) is expressed in a wide range of epithelial structures in humans. CK7 occurs in the columnar and glandular epithelium of the lug, cervix, breast, in bile ducts, collecting ducts in the kidney and mesothelium, but is absent in gastrointestinal epithelium, hepatocytes, proximal and distal tubules of the kidney and myoepithelium. The antibody OVTL 12/30 is an excellent tool in the differential diagnosis of malignant cells and to determine primary tumor localization. It stains a 54kD protein band of cytoskeletal proteins from cell lines in one and two dimensional immunoblots.
Catalog No. 50-238-5594
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.