Learn More
Biocare Medical, LLC Cytokeratin 7 (CK7) Antibody - Predilute, 6.0 ml
SDP
Supplier:  Biocare Medical, LLC PM061AA
Encompass_Preferred
CK7 antibody is an intermediate filament protein (IFP) of 54 kDa that recognizes the simple epithelium found in most glandular and transitional epithelia; but not in stratified squamous epithelia. This monoclonal antibody [OV-TL 12/30] has been shown to be highly specific to Cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin and is expressed in epithelial cells of ovary, lung and breast, but not of colon or gastrointestinal tract. It is often used in concert with Cytokeratin 20 and CDX-2 to aid in distinguishing ovarian, pulmonary and breast carcinomas (CK7 ) from colon carcinomas (CK7-).
Catalog No. 50-830-26
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.