Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 7 Monoclonal Antibody (OV-TL12/30)
GREENER_CHOICE

Catalog No. 180234
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-023-4 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-023-4 Supplier Invitrogen™ Supplier No. 180234
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

The 54 kDa protein, cytokeratin 7, is found in most glandular and transitional epithelia. It has detected cytokeratin 7 in a large number of epithelial cell types, including many ductal and glandular epithelia by immunohistochemistry of paraffin-embedded tissue. Primarily, cytokeratin 7 antibody does not react with stratified squamous epithelia but is reactive with transitional epithelium of the urinary tract. Positive tissue: Pancreas/bladder cancer. This antibody is specific for cytokeratin 7, and does not exhibit cross-reactivity with other filament types, including vimentin, desmin, glial fibrillary acidic protein and neurofilament protein. Expected Staining Pattern: Cytoplasmic.

Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 7
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OV-TL12/30
Concentration 0.03755 mg/mL
Conjugate Unconjugated
Dilution Concentrate Antibody
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene KRT7
Gene Accession No. P08729
Gene Alias CK7; CK-7; Cytokeratin 7; cytokeratin-7; D15Wsu77e; K2C7; K7; Kb7; keratin 7; keratin 7, type II; keratin associated protein 7.1; keratin complex 2, basic, gene 7; keratin, 55K type II cytoskeletal; keratin, simple epithelial type I, K7; keratin, type II cytoskeletal 7; Keratin-7; keratin-associated protein 7-1; keratin-associated protein 7-1-like protein; Krt2-7; KRT7; krt7 {ECO:0000250; LOC100755511; LOC100861181; NACHT, LRR and PYD domains-containing protein 3; sarcolectin; SCL; si:ch73-233m11.1; si:ch73-233m11.2; similar to keratin 7; type II keratin Kb7; type II mesothelial keratin K7; type-II keratin Kb7; UniProtKB:P08729}
Gene Symbols KRT7
Host Species Mouse
Immunogen OTN11 human ovarian carcinoma cells.
Molecular Weight of Antigen 54kDa
Positive Control Pancreas/Bladder cancer
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Packaging 1 Each
Test Specificity This antibody is specific for cytokeratin 7, and does not exhibit cross-reactivity with other filament types, including vimentin, desmin, glial fibrillary acidic protein and neurofilament protein
Includes 0.1% Sodium azide
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Reactivity Human
Shipping Condition Wet Ice
Reactivity abrv Hu
Target Gene Cytokeratin 7
Validated Application abrv IHC (FFPE)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.