Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytokeratin 7 Mouse anti-Human, Clone: 2E1G5, Proteintech
SDP

Catalog No. 501736906 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6906 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6906 Supplier Proteintech Group Inc Supplier No. 664831IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody is specifically against KRT7.

Cytokeratin 7 blocks interferon-dependent interphase and stimulates DNA synthesis in cells. Involved in the translational regulation of the human papillomavirus type 16 E7 mRNA (HPV16 E7).

Specifications

Antigen Cytokeratin 7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2E1G5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene KRT7
Gene Accession No. P08729
Gene Alias CK 7, CK7, Cytokeratin 7, K2C7, K7, keratin 7, KRT7, Sarcolectin, SCL, Type II keratin Kb7
Gene Symbols KRT7
Host Species Mouse
Immunogen Cytokeratin 7 Fusion Protein ag7895
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3855
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.