Learn More
Description
Host Species: Mouse
Clone: 5D3
Isotype: IgG2a
Species Reactivity: Human. Others not known.
Immunogen: Cytokeratins purified from breast cancer MCF-7 cells
Molecular Weight: 52.5 kDa (Cytokeratin 8) and 45 kDa (Cytokeratin 18)
Positive Control: Skin
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Cytokeratin 8/18 Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 5D3 |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Cytokeratins purified from breast cancer MCF-7 cells |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Research Discipline | Cytoskeletal Protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.