Learn More
Carbosynth LLC. Guinea Pig polyclonal Cytokeratin 8 +18 antibody
SDP
Supplier:  Carbosynth LLC. 20RCP004
Encompass_Preferred
Cytokeratins are proteins of keratin-containing intermediate filaments found in the intracytoplasmic cytoskeleton of epithelial tissue. Cytokeratin represents an excellent marker for the discrimination of specific subtypes of adenocarcinoma. Keratin, type I cytoskeletal 8 also known as cytokeratin-8 (CK-8) or keratin-8 (K8) is a keratin protein that in human is encoded by the KRT8 gene. It is often paired with keratin 18.
Catalog No. 50-125-2887
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.