Learn More
Description
MA5-14088 targets Cytokeratin 8/18 in immunohistochemistry (paraffin) applications and shows reactivity with Human samples. The MA5-14088 immunogen is cytokeratins purified from breast cancer MCF-7 cells.
Specifications
Specifications
| Antigen | Cytokeratin 8/18 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 5D3 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.09% sodium azide |
| Gene | KRT8 |
| Gene Accession No. | P05783, P05787 |
| Gene Alias | AA960620; AL022697; AU019895; CARD2; cell proliferation-inducing gene 46 protein; CK18; CK-18; CK8; CK-8; CYK18; CYK8; cytokeratin 18; cytokeratin 8; cytokeratin endo A; Cytokeratin endo B; cytokeratin-18; Cytokeratin-8; Endo B; EndoA; K18; K2C8; K8; keratin 18; keratin 18, type I; keratin 8; keratin 8, type II; keratin complex 1, acidic, gene 18; keratin complex 2, basic, gene 8; keratin D; Keratin, type I cytoskeletal 18; keratin, type II cytoskeletal 8; Keratin-18; keratin-8; Kerd; KO; Krt1-18; KRT18; Krt-2.8; Krt2-8; Krt8; PIG46; Type-II keratin Kb8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
