Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytokeratin 8/18 Monoclonal Antibody (Zym5.2 (UCD/PR.10-11))
GREENER_CHOICE

Catalog No. 180213
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-021-3 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-021-3 Supplier Invitrogen™ Supplier No. 180213
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This monoclonal antibody reacts with human cytokeratins 8 and 18, low molecular weight cytokeratins found in simple epithelium and secretory epithelium. Clone Zym5.2 will react with adenocarcinoma, mesothelioma, and carcinoma derived from simple epithelia, but will not react with stratified squamous epithelia. Positive tissue: Normal colon.Clone/PAD: Zym5.2 (UCD/PR.10-11)Immunogen: Human keratin 8 & 18Specificity: Cytokeratin 8 (52.5 kDa) and 18 (45 kDa) Expected Staining Pattern: Cytoplasmic and membrane.

Keratin 8 belongs to the type B (basic) subfamily of high molecular weight keratins and exists in combination with cytokeratin 18 (type A; acidic subfamily of low molecular weight keratins).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytokeratin 8/18
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone Zym5.2 (UCD/PR.10-11)
Concentration 0.34443 mg/mL
Conjugate Unconjugated
Dilution Concentrate Antibody
Formulation PBS with 1% BSA and 0.1% sodium azide
Gene KRT8
Gene Accession No. P05783, P05787
Gene Alias AA960620; AL022697; AU019895; CARD2; cell proliferation-inducing gene 46 protein; CK18; CK-18; CK8; CK-8; CYK18; CYK8; cytokeratin 18; cytokeratin 8; cytokeratin endo A; Cytokeratin endo B; cytokeratin-18; Cytokeratin-8; Endo B; EndoA; K18; K2C8; K8; keratin 18; keratin 18, type I; keratin 8; keratin 8, type II; keratin complex 1, acidic, gene 18; keratin complex 2, basic, gene 8; keratin D; Keratin, type I cytoskeletal 18; keratin, type II cytoskeletal 8; Keratin-18; keratin-8; Kerd; KO; Krt1-18; KRT18; Krt-2.8; Krt2-8; Krt8; PIG46; Type-II keratin Kb8
Gene Symbols KRT18, KRT8
Host Species Mouse
Immunogen Human keratin 8 & 18.
Positive Control Normal colon
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3856, 3875
Packaging 1 Each
Test Specificity Cytokeratin 8 (52.5kDa) and 18 (45kDa)
Includes 0.1% Sodium azide
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Isotype IgG1
Host abrv Ms
Reactivity Human
Shipping Condition Wet Ice
Reactivity abrv Hu
Target Gene Cytokeratin 8/18
Validated Application abrv IHC (FFPE)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.