Learn More
Diagnostic Biosystems CYTOKERATIN 8 Cytokeratin 8, Low Molecular Weight
Supplier:  Diagnostic Biosystems PDM117
Encompass_Preferred
This antibody recognizes keratin polypeptide of 54 kDa. It stains strongly with non-squamous epithelium but does not stain squamous epithelium. It reacts with the sweat gland and ducts in the skin. Adenocarcinomas of the ovary, gastrointestinal tract, ductal carcinoma of breast, pancreas and bile ducts, stain positive with this antibody.
Catalog No. 50-238-5334
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.