Learn More
Biogenex Laboratories Cytokeratin Cocktail : 6 mL
SDP
Supplier:  Biogenex Laboratories AM3725M
Encompass_Preferred
Human cytokeratins, a family of water-insoluble polypeptides, form themajor part of the cytoskeleton in all normal and neoplastic epithelial cells.CK88 is a broad spectrum antibody cocktail that reacts with a variety ofnormal and neoplastic epithelia. It recognizes most epithelium includingsimple, basal, suprabasal layers, cornea, cornifying stratified epithelium ofskin, transitional epithelium of urinary tract, and squamous epithelium.Analysis of intracellular keratin by immunoperoxidase technique is helpfulin establishing the epithelial nature of primary or metastatic poorlydifferentiated neoplasms.
Catalog No. 50-197-8849
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.