Learn More
Cytokeratin, pan Antibody (AE-1/AE-3), Alexa Fluor™ 647, R&D Systems™

Description
Specifications
Specifications
| Antigen | pan Cytokeratin |
| Applications | CyTOF, Dual RNAScope ISH-IHC, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Single Cell Western, Western Blot |
| Classification | Monoclonal |
| Clone | AE-1/AE-3 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular) |
| Gene Symbols | KRT1 |
| Host Species | Mouse |
| Immunogen | This Cytokeratin, pan Antibody (AE-1/AE-3) was developed against total keratin isolated from human epidermal callus was used as immunogen to generate the pan cytokeratin antibodies AE1 + AE3 (Woodcock-Mitchell, 1982). |
| Purification Method | Protein A or G purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.