Learn More
Sartorius Incucyte Cytolight Rapid Dye for Live-cell Cytoplasmic Labeling, Red
Catalog No. NC1353581 Shop All Sartorius Products
Encompass
Supplied as dry powder in sufficient quantity capable of labeling between 10 and 90 million cells depending on the optimized concentration.
Catalog No. NC1353581 Supplier Sartorius Supplier No. 4706
Supplied as dry powder in sufficient quantity capable of labeling between 10 and 90 million cells depending on the optimized concentration.