Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cytomegalovirus IE 72 kDa Antigen Monoclonal Antibody (1K10)

Catalog No. 01670330 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-670-330 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-670-330 Supplier Invitrogen™ Supplier No. MA17596
  • $598.00
    $489.65 / Each of 1

    Save $108.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA1-7596 detects HCMV from virus samples. The MA1-7596 immunogen is an HCMV IE concentrate. Store short-term at 4°C, long-term at -80°C.

Cytomegalovirus (CMV) is an opportunistic pathogen infecting lung, kidney, gut and other organs in situations where an individual is immunologically immature, such as the foetus and neonate. Infection also occurs in immunosuppressed patients e.g. transplant patients, patients undergoing chemotherapy and those with HIV infection. Cytomegalovirus is an enveloped double stranded DNA virus which in humans (HCMV) is commonly known as human herpesvirus 5 (HHV-5).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cytomegalovirus IE 72 kDa Antigen
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1K10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.01% sodium azide; pH 7.2
Gene Alias CMV
Host Species Mouse
Immunogen HCMV IE concentrate.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage 4°C short term, -80°C long term
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.