Learn More
Genscript Corporation CytoSinct CD8 Nanobeads huma
Supplier:  Genscript Corporation L009337.5
Encompass_Preferred
The CytoSinct CD8 Nanobeads human (GMP) is used for in vitro isolation or depletion CD8 cells from fresh or frozen peripheral blood mononuclear cells (PBMCs) leukapheresis products or single cell suspension based on the surface expression of human CD8 Due to the nanoscale structure of the CytoSinct CD8 Nanobeads human (GMP) the excess reagent can be removed by centrifugation or conventional supernatant replacement
Catalog No. 50-000-59050
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.