Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytosine, MP Biomedicals™

Catalog No. ICN10146901 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN10146901 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10146901 Supplier MP Biomedicals, Inc Supplier No. 0210146901

missing translation for 'validMainframeMsg'

Cytosine (C) is one of the four main bases found in DNA and RNA, along with adenine, guanine, and thymine (uracil in RNA)

TRUSTED_SUSTAINABILITY

Specifications

CAS 71-30-7
Color Yellow
Molecular Formula C4H5N3O
Quantity 1 g
MDL Number MFCD00006034
Synonym cytosine, 4-amino-2-hydroxypyrimidine, cytosinimine, 2 1h-pyrimidinone, 4-amino, 4-amino-2 1h-pyrimidinone, 4-aminopyrimidin-2 1h-one, 6-aminopyrimidin-2 1h-one, 2 1h-pyrimidinone, 6-amino, cyt, 4-amino-2-oxo-1,2-dihydropyrimidine
InChI Key OPTASPLRGRRNAP-UHFFFAOYSA-N
SMILES NC1=CC=NC(=O)N1
IUPAC Name 6-amino-1,2-dihydropyrimidin-2-one
Molecular Weight (g/mol) 111.10
PubChem CID 597
ChEBI CHEBI:16040
Formula Weight 111.1
Physical Form Powder
Chemical Name or Material Cytosine, FTIR
Show More Show Less
EINECSNumber 200-749-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.