Learn More
Advanced Biomatrix CytoSoft Imaging 96-well Plate, Elastic Modulus 64 kPa
Supplier:  Advanced Biomatrix 52611EA
Encompass_Preferred
This CytoSoft Imaging 96-well product has a defined elastic modulus (see certificate of analysis) with a glass bottom for high resolution imaging The thickness of the silicone gel is uniform with a ~0.03 mm thick layer of silicone in each well. The silicone gels are activated and ready to bind to a purified ECM.The rigidity of the substrate to which cells adhere can have a profound effect on cell morphology and gene expression. The CytoSoft Imaging plate has a glass bottom for high-resolution imaging where low autofluorescence and exceptional optical clarity are required. The 96-well format is optimal for high-throughput cell studies and screening. On the bottom of each well, there is a thin layer of specially formulated biocompatible silicone, whose elastic modulus (rigidity) is carefully measured and certified.
Catalog No. 50-196-0305
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.