Description
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
- Clipped Cytofunnel Starter Kit
- Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
- EZ Cytofunnel Starter Kit
- Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
Specifications
Specifications
| For Use With (Equipment) | Cytospin™ 4 Cytocentrifuge |
| Product Type | Clipped Funnel Starter Kit |