Learn More
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit
GSA_VA
Catalog No. p-9397008 Shop All Epredia Products
Change view
Click to view available options
Product Type:
Clipped Funnel Starter Kit
EZ Cytofunnel Starter Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
A78710013 EZ Cytofunnel Starter Kit
31-201-10 Clipped Funnel Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. A78710013 Supplier Epredia™ Supplier No. A78710013
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Accessory Starter Kit for the Epredia™ Cytospin™ 4 Cytocentrifuge. (Centrifuge is sold separately, Cat. No. A78300003)
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
  • Clipped Cytofunnel Starter Kit
    • Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
  • EZ Cytofunnel Starter Kit
    • Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
For Use With (Equipment) Cytospin™ 4 Cytocentrifuge
Product Line Cytospin
Frequency 50/60 Hz
Voltage 100/240 V
Product Type EZ Cytofunnel Starter Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov