Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-2-(2,5-Dihydrophenyl)glycine 97.0+%, TCI America™
SDP

Catalog No. D253925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D253925G 25 g
D25395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D253925G Supplier TCI America Supplier No. D253925G

Chemical Identifiers

CAS 26774-88-9
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.181
MDL Number MFCD00137746
InChI Key JBJJTCGQCRGNOL-SSDOTTSWSA-N
Synonym r-2-amino-2-cyclohexa-1,4-dien-1-yl acetic acid, r---2-2,5-dihydrophenyl glycine, unii-451nq27tg5, 2,5-dihydrophenylglycine, d, r---2-amino-2-cyclohexa-1,4-dienyl acetic acid, d---2-2,5-dihydrophenyl glycine, r-alpha-amino-1,4-cyclohexadiene-1-acetic acid, 2r-2-amino-2-cyclohexa-1,4-dienylacetic acid, r-amino cyclohexa-1,4-dien-1-yl acetic acid, d-cyclohexadienylglycine
PubChem CID 1268102
IUPAC Name (2R)-2-amino-2-cyclohexa-1,4-dien-1-ylacetic acid
SMILES C1C=CCC(=C1)C(C(=O)O)N

Specifications

Quantity 25 g
Formula Weight 153.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-2-(2,5-Dihydrophenyl)glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.