Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(-)-2-Phenylglycine chloride hydrochloride, 98%, Thermo Scientific™

Catalog No. AAH5682803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5682803 1 g
AAH5682806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5682803 Supplier Thermo Scientific Chemicals Supplier No. H5682803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39878-87-0
Molecular Formula C8H9Cl2NO
Molecular Weight (g/mol) 206.066
MDL Number MFCD00039094
InChI Key GVVFCAFBYHYGEE-OGFXRTJISA-N
Synonym r-2-amino-2-phenylacetyl chloride hydrochloride, r---2-phenylglycine chloride hydrochloride, unii-07v2dd878h, 2r-2-amino-2-phenylacetyl chloride hydrochloride, --alpha-chloroformyl benzylammonium chloride, benzeneacetyl chloride, alpha-amino-, hydrochloride, alphar, d---phenylglycine acid chloride, hydrochloride, r-alpha-aminobenzeneacetyl chloride, hydrochloride, r---2-phenylglycine chloride hcl, 2r-amino phenyl acetoyl chloride hydrochloride 1:1
PubChem CID 170253
IUPAC Name (2R)-2-amino-2-phenylacetyl chloride;hydrochloride
SMILES C1=CC=C(C=C1)C(C(=O)Cl)N.Cl

Specifications

Melting Point 177°C
Color Brown
UN Number UN3261
Beilstein 5405183
Quantity 1 g
Flash Point 150°C (302°F)
Formula Weight 206.07
Sensitivity Moisture sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-(-)-2-Phenylglycine chloride hydrochloride
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 254-668-5
RTECSNumber CY1920000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.