Learn More
Medchemexpress LLC HY-41700 500mg Medchemexpress, D-Alanine CAS:338-69-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY41700500MG
Encompass_Preferred
Medchemexpress, HY-41700 500mg D-Alanine CAS:338-69-2 D-Alanine is a weak GlyR (inhibitory glycine receptor) and PMBA agonist, with an EC50 of 9 mM for GlyR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2239
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.