Learn More
D-Alanine methyl ester hydrochloride, 98%
Description
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
D-Alanine methyl ester hydrochloride is used as building block for the preparation of peptides.
Solubility
Soluble in Water 100 mg/ml.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Specifications
Specifications
| Melting Point | 108°C to 110°C |
| Color | White |
| Beilstein | 4235975 |
| Solubility Information | Soluble in Water 100mg/ml. |
| Quantity | 5 g |
| Formula Weight | 139.58 |
| Percent Purity | 98% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | D-Alanine methyl ester hydrochloride |
Safety and Handling
| EINECSNumber | 238-258-3 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.