Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-alpha-Tocopherol, 97+%

Catalog No. AC421031000
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421031000 100 mg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC421031000 Supplier Thermo Scientific Chemicals Supplier No. 421031000
  • $212.00
    $169.65 / Each of 1

    Save $42.35

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59-02-9 | C29H50O2 | 430.7 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-02-9
Molecular Formula C29H50O2
Molecular Weight (g/mol) 430.7
MDL Number MFCD00072045
InChI Key GVJHHUAWPYXKBD-IEOSBIPESA-N
Synonym vitamin e, alpha-tocopherol, d-alpha-tocopherol, 5,7,8-trimethyltocol, +-alpha-tocopherol, r,r,r-alpha-tocopherol, phytogermine, eprolin, 2r,4'r,8'r-alpha-tocopherol, dl-a-tocopherol
PubChem CID 14985
ChEBI CHEBI:18145
IUPAC Name (2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC1=C(C(=C2CCC(OC2=C1C)(C)CCCC(C)CCCC(C)CCCC(C)C)C)O

Specifications

Melting Point 2.5°C to 3.5°C
Density 0.9500g/mL
Boiling Point 200°C to 220°C (0.1 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5050 to 1.507
Quantity 100 mg
Specific Gravity 0.95
Merck Index 15, 9653
Specific Rotation Condition +24° (25°C c=2,in isooctane)
Specific Rotation + 24
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,chloroform,ether,,oils,fats
Formula Weight 430.7
Percent Purity 97+%
Physical Form Viscous Liquid
Chemical Name or Material D-α-Tocopherol, 99%
Show More Show Less
Hazard Category LIGHT SENSITIVE
Hazard Statement AIR SENSITIVE
EINECSNumber 200-412-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.