Learn More
Sigma Organic Chemistry D-alpha-Tocopherylquino | 50mg | MFCD00135657
Supplier: Sigma Organic Chemistry CDS00003850MG

D-alpha-Tocopherylquino , 50mg
About this Item:
MDL #: MFCD00135657
Molecular Weight: 446.71
UNSPSC Code: 12352200
Chemical Formula: C29H50O3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.