Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ D-AP4

Catalog No. 010210
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-021-0 10 mg
01-025-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 01-021-0 Supplier Tocris Bioscience™ Supplier No. 0102/10
This item is not returnable. View return policy

Broad spectrum glutamatergic antagonist

D-AP4 is a broad spectrum excitatory amino acid receptor antagonist. Also agonist at the quisqualate-sensitized AP6 site in hippocampus where it is less potent than L-AP4.

DL Mixture and L-isomer also available.

Specifications

Chemical Name or Material D-(-)-2-Amino-4-phosphonobutyric acid
CAS 78739-01-2
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 100 mM in 1eq. NaOH
Molecular Formula C4H10NO5P
Formula Weight Observed MW: 183.1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.