Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Arabinitol, MP Biomedicals

Catalog No. ICN222436 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN222436 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN222436 Supplier MP Biomedicals, Inc Supplier No. 0522243601

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 488-82-4
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.146
InChI Key HEBKCHPVOIAQTA-QWWZWVQMSA-N
Synonym d-arabinitol, arabitol, dl-arabitol, d-arabitol, d-+-arabitol, d-+-arabinitol, d-lyxitol, lyxitol, dl-arabinitol, d-arabinol
PubChem CID 94154
ChEBI CHEBI:18333
IUPAC Name (2R,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

Quantity 1 g
Formula Weight 152.1
Chemical Name or Material D-(+)-Arabinitol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.