Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Arabinose, MP Biomedicals™

Catalog No. ICN19402380 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19402380 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19402380 Supplier MP Biomedicals, Inc Supplier No. 0219402380
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10323-20-3
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key SRBFZHDQGSBBOR-UHFFFAOYNA-N
Synonym d---arabinose, d-arabinose, dl-arabinose, arabinose, d, arabinose,d, 2s,3r,4r-2,3,4,5-tetrahydroxypentanal, unii-f0w6etz4e5, --arabinose, aldehydo-d-arabinose, f0w6etz4e5
PubChem CID 66308
ChEBI CHEBI:46983

Specifications

Quantity 100 g
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.