Learn More
Medchemexpress LLC HY-W016781 500mg Medchemexpress, D-Arginine CAS:157-06-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW016781500MG
Encompass_Preferred
Medchemexpress, HY-W016781 500mg D-Arginine CAS:157-06-2 D-arginine (H-D-Arg-OH) is the D-isomer of arginine. Arginine is an α-amino acid that is used in the biosynthesis of proteins. D-Arginine is an inactive form of L-arginine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3400
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.