Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-beta-Proline, 98+%

Catalog No. AAH5735603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5735603 1 g
AAH57356MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5735603 Supplier Thermo Scientific Chemicals Supplier No. H5735603
Add to Cart

CAS: 72580-54-2 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72580-54-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD08272856
InChI Key JAEIBKXSIXOLOL-SCSAIBSYSA-N
Synonym r-pyrrolidine-3-carboxylic acid, 3r-pyrrolidine-3-carboxylic acid, r---pyrrolidine-3-carboxylic acid, d-beta-proline, r-beta-proline, 3-pyrrolidinecarboxylic acid, 3r, r-3-pyrrolidinecarboxylic acid, l-pyrrolidine-3-carboxylic acid, 3r-3-pyrrolidinecarboxylic acid hcl, r-pyrrilidine-3-carboxylic acid
PubChem CID 1501970
IUPAC Name (3R)-pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 185°C to 191°C
Beilstein 5496144
Quantity 1 g
Formula Weight 115.13
Optical Rotation −18° (c=2 in Water)
Percent Purity ≥98%
Chemical Name or Material D-beta-Proline
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.