Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ D-Biotin

Catalog No. B1595
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1595 1 g
B20656 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1595 Supplier Invitrogen™ Supplier No. B1595
Add to Cart
Add to Cart

D-Biotin

D-biotin binds with high affinity to streptavidin, avidin and NeutrAvidin™ biotin-binding proteins. This solution can be used in conjunction with unlabeled streptavidin or avidin in procedures to block endogenous biotin.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Unconjugated
Quantity 1 g
Content And Storage Store at room temperature.
Shipping Condition Room Temperature
Form Solid
Product Line Invitrogen
Product Type Biotin (unconjugated)
How should I resolubilize D-biotin (B20656)?

D-biotin may be resolubilized in DMF (1-2 mg/mL), DMSO (49 mg/mL), ethanol (<1 mg/mL) or a buffer with a pH between >6 and <9. For long-term stability, D-biotin is stable in aqueous buffers at moderately acidic to neutral pH.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.