Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-Biotin, ≥97.5%, Cell Culture Reagent Grade, MP Biomedicals™

Catalog No. ICN19463405 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19463405 5 g
ICN19463401 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19463405 Supplier MP Biomedicals, Inc Supplier No. 0219463405
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 58-85-5
Molecular Formula C10H16N2O3S
Molecular Weight (g/mol) 244.31
MDL Number MFCD00005541
InChI Key YBJHBAHKTGYVGT-UHFFFAOYNA-N
Synonym biotin, d-biotin, vitamin h, coenzyme r, vitamin b7, bios ii, factor s, bioepiderm, d +-biotin, biodermatin
PubChem CID 171548
ChEBI CHEBI:15956
SMILES OC(=O)CCCCC1SCC2NC(=O)NC12

Specifications

Color White
Density 1.267 g/cm3(Lit.)
Assay Percent Range ≥97.5%
Quantity 5 g
IUPAC Name 5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanoic acid
Formula Weight 244.3
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material D-Biotin
EINECSNumber 200-399-3
Recommended Storage Store at +4°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.