Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Green No. 5, Spectrum™ Chemical
SDP

Catalog No. 18601969 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-969 500 g Amber Glass Bottle
18-601-967 25 g Amber Glass Bottle
18-601-968 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-969 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1119500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

D1119, 4403-90-1, C28H20N2O8S2Na2

Spectrum™ Chemical D and C Green No. 5

Chemical Identifiers

CAS 4403-90-1
Molecular Formula C28H20N2Na2O8S2
Molecular Weight (g/mol) 622.57
InChI Key FPAYXBWMYIMERV-UHFFFAOYSA-L
IUPAC Name disodium 5-methyl-2-({4-[(4-methyl-2-sulfonatophenyl)amino]-9,10-dioxo-9,10-dihydroanthracen-1-yl}amino)benzene-1-sulfonate
SMILES [Na+].[Na+].CC1=CC=C(NC2=CC=C(NC3=CC=C(C)C=C3S([O-])(=O)=O)C3=C2C(=O)C2=CC=CC=C2C3=O)C(=C1)S([O-])(=O)=O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Color Blue/Green
Quantity 500 g
Grade Reagent
Chemical Name or Material D and C Green No. 5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.