Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D and C Red No. 33, Spectrum™ Chemical
SDP

Catalog No. 18601979 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-979 25 g Amber Glass Bottle
18-601-980 100 g Amber Glass Bottle
18-601-981 500 g Amber Glass Bottle
18-601-982 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-979 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. DC14525GM
Add to Cart
Add to Cart

DC145, 3567-66-6, C16H11N3O7S2Na2

Spectrum™ Chemical D and C Red No. 33

Chemical Identifiers

CAS 3567-66-6

Specifications

CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Color Red
Quantity 25 g
Chemical Name or Material D and C Red No. 33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.