Learn More
United Scientific D Cell Battery Holder and Joiner
SDPSCI_ED
Catalog No. S81068 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
For Use With (Application):
Electricity Demonstration
Catalog No. For Use With (Application)
S81068 Electricity Demonstration
1 options
Catalog No. S81068 Supplier United Scientific Supplier No. BTHD01
Add to Cart
Add to Cart
Battery Holder for use with a variety of electricity demonstrations.
  • Single holders can be snapped together in parallel or series to extend capacity or voltage
  • Includes male and female button snaps on ends to allow series connections and includes extensions on sides for parallel connections
  • Prop 65 Warning Label
Specifications
For Use With (Application) Electricity Demonstration
Product Type Battery Holder