Learn More
Carbosynth LLC. D-Cellobiose, 528-50-7, MFCD00136034, 100 g
Supplier: Carbosynth LLC. OC04040100G
This item has a minimum order quantity of 2 per supplier requirements
Formula: C12H22O11, Molecular weight: 342.30 g/mol, 98%, Store at room temperature, Synonyms: 4-O-beta-D-Glucopyranosyl-D-glucose.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.