Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Cellobiose; ≥99%, MP Biomedicals™

Catalog No. ICN10129880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10129880 100 g
ICN10129805 5 g
ICN10129810 10 g
ICN10129825 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10129880 Supplier MP Biomedicals, Inc Supplier No. 0210129880
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cellobiose is a disaccharide composed of two glucose molecules; These are β-1,4-glycosidically linked. It is reducing sugar, which is readily soluble in polar solvents such as water and ethanol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range ≥99%
Quantity 100 g
IUPAC Name 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
Formula Weight 342.3
Percent Purity ≥99%
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
EINECSNumber 208-436-5
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.