CAS RN 528-50-7
Thermo Scientific Chemicals D(+)-Cellobiose, 98%
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 IUPAC Name: (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
Thermo Scientific Chemicals D-(+)-Cellobiose, 98+%
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
D(+)-Cellobiose; ≥99%, MP Biomedicals™
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
D(+)-Cellobiose; 99%, MP Biomedicals™
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 IUPAC Name: (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
D-(+)-Cellobiose, MP Biomedicals™
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 IUPAC Name: (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O
MilliporeSigma™ Calbiochem™ D(+)Cellobiose
Substrate for β-glucosidase
D-(+)-Cellobiose 98.0+%, TCI America™
CAS: 528-50-7 Molecular Formula: C12H22O11 Molecular Weight (g/mol): 342.30 MDL Number: MFCD00136034 InChI Key: GUBGYTABKSRVRQ-UHFFFAOYNA-N Synonym: d-+-cellobiose,4-o-beta-glucopyranosyl-d-glucose PubChem CID: 57370084 IUPAC Name: 2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol SMILES: OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O