Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D(+)-Cellobiose; 99%, MP Biomedicals™

Catalog No. ICN10031910 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10031910 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10031910 Supplier MP Biomedicals, Inc Supplier No. 0210031910
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Major electron acceptor molecule in biological oxidations that acts as a coenzyme for hydride-transfer enzymes.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range 0.99
Quantity 10 g
Formula Weight 342.3
Percent Purity 93%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Cellobiose
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.