Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

D-(+)-Cellobiose, MP Biomedicals™

Catalog No. MP210129890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP210129890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210129890 Supplier MP Biomedicals, Inc Supplier No. 0210129890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Readily soluble in polar solvents, such as water and ethanol. D-(+)-Cellobiose, MP Biomedicals is a disaccharide composed of two glucose molecules. It is a reducing sugar that is used as a cell culture media supplement. These are β-1,4-glycosidically

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 528-50-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00136034
InChI Key GUBGYTABKSRVRQ-UHFFFAOYNA-N
Synonym d-+-cellobiose, 4-o-beta-glucopyranosyl-d-glucose
PubChem CID 57370084
IUPAC Name (3S,4S,6S)-2-(hydroxymethyl)-6-[(3S,4R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol
SMILES OCC1OC(OC2C(O)C(O)C(O)OC2CO)C(O)C(O)C1O

Spécifications

Melting Point 229°C to 239°C
Quantity 500 g
Solubility Information Soluble in water 4% (clear colorless solution); insoluble in alcohol and ether.
Optical Rotation +34° ± 3° (c=4, water)
Formula Weight 342.3
Percent Purity ≥99%
Grade Not Available
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'

missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.