Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals D-(+)-Cellotetraose, 95%

Catalog No. AAJ67583MA
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67583MA 10 mg
AAJ67583LB0 5 mg
AAJ67583MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ67583MA Supplier Thermo Scientific Chemicals Supplier No. J67583MA
Add to Cart

CAS: 38819-01-1 | C24H42O21 | 666.579 g/mol

Derived from cellulose degradation. Used by cellulosic bacteria as an energy source. Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and --glucosidases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Derived from cellulose degradation
  • Used by cellulosic bacteria as an energy source
  • Useful for characterization and identification of oligosaccharide metabolizing enzymes such as endoglucanases and _x001A_-glucosidases
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38819-01-1
Molecular Formula C24H42O21
Molecular Weight (g/mol) 666.579
MDL Number MFCD00079079
InChI Key LUEWUZLMQUOBSB-HQXNUCTASA-N
Synonym cellotetraose
PubChem CID 131674892
IUPAC Name (2S,3S,5S)-2-[(3S,5S,6S)-6-[(3S,5S,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(3S,5S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)OC4C(OC(C(C4O)O)O)CO)CO)CO)O)O)O)O

Specifications

Packaging Plastic bottle
Quantity 10 mg
Beilstein 78869
Sensitivity Hygroscopic
Solubility Information Soluble at 25mg/ml in water
Formula Weight 666.58
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-(+)-Cellotetraose
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.